Clips for conical joints
Supplier: Roth Carl
1163.1 1164.1 1165.1 1166.1 1167.1 1168.1 1169.1
ROTH1163.1EA 0 EUR
ROTH1163.1 ROTH1164.1 ROTH1165.1 ROTH1166.1 ROTH1167.1 ROTH1168.1 ROTH1169.1
Clips for conical joints
Clamps NS Joint Clamps
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
Delivery information: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).