Tubing, rubber, Rotilabo®
Supplier: Roth Carl
T981.1 T982.1 T983.1 T984.1 0674.1 0675.1 0676.1 0677.1
ROTHT981.1EA 64.1 EUR
ROTHT981.1 ROTHT982.1 ROTHT983.1 ROTHT984.1 ROTH0674.1 ROTH0675.1 ROTH0676.1 ROTH0677.1
Tubing, rubber, Rotilabo®
Tubing Transfer Tubing
Natural rubber (NR), red.
  • Temperature resistance −10 to +50 °C
  • Not pressure resistant
  • Hardness: 45 Shore A