MµltiGuard™ Barrier Pipette Tips, Sterile
Supplier: Sorenson Bioscience
LAC10420 LAC10400 LAC10460 38000T 14200
613-3049EA 167 EUR
613-3049 613-3047 613-3051 613-3031 SORE14200
MµltiGuard™ Barrier Pipette Tips, Sterile
Pipette Tips
Standard tips for general pipetting applications. Designed to fit most research-grade pipettors.
  • Available with low binding surface technology for reduced sample retention
  • Certified free from RNases, DNases, human DNA, PCR inhibitors and pyrogens
  • XT tips (extended length): Ideal for pipetting from deep vessels
  • LongReach, NX and type E tips: Ideal for molecular biology applications like PCR
  • OneTouch tips create a leak proof seal: Easy to seal, easy to eject
Packaging options:
Racked
Sturdy racks that withstand several autoclave cycles
Cell Racks hold tips steadily in fully enclosed cells, minimise static and are excellent platforms for single and multichannel pipetting
Hinged racks hold the extremely popular 1000XT (50 to 1250 µl) tips, and traditional 1000 µl standard tips are placed in 100-hole standard racks
e•dek – Paperboard Rack System
Easy to recycle – compatible with most paperboard recycling programs
Used racks require minimal storage space – just flatten and recycle
Manufactured from paperboard sourced from sustainable forests
Printed with soy and vegetable based inks
Less weight equals reduced energy consumption and cost for shipping
Informace pro objednávání: Supplied in polypropylene CellRacks. MµltiGuard™ tips 14234-104 and 14234-106 can accommodate a volume of 30 µl.
Balení: Packs contain 10 racks with 96 tips/rack, except 14234-120 and 14234-122, which have 100 tips/rack.