Large swab rinse kits, SRK®
Supplier: Copan
904C 905C
710-0474EA 41.6 EUR
710-0474 710-0475
Large swab rinse kits, SRK®
Swabs Swabs with Rinse Solutions
Large swab rinse kits (SRK®) for microbiological examination of surfaces.
  • Isotonic salt solution with substances to neutralise and deactivate disinfectants and sanitising agents
  • Rinse solution to moisten the sterile swab before sampling and to transport the swab after collection
  • Swabs are separate from cap
Suitable for microbial detection or quantitative analysis.
Balení: Individually wrapped in plastic peel pouch.