Disinfectant wipes, Kohrsolin® FF Tissues
Supplier: Bode
Kohrsolin® FF
BODE9812011EA 0 EUR
BODE9812011 115-1384
Disinfectant wipes, Kohrsolin® FF Tissues
Wipes Disposable Wipes and Towels Disinfectant Wipes
These aldehydes containing fleece wipes are suitable for daily prophylactic disinfectant cleaning of washable surfaces of medical devices and equipment (in accordance with MPG), as well as in hospitals, doctors' surgeries and nursing homes (in accordance with BPD).
  • Virucidal activity against murine noroviruses
  • Good material compatibility
  • Good wetting and cleaning, no adsorption
  • High quality, soft wipe fabric
  • Safe and easy dispensing of individual wipes from resealable pack
The wipes leave very little residue and are effective against murine noroviruses (MNV), a surrogate virus for noroviruses that has not yet been recognised by the RKI.
Microbiological effectiveness: bactericidal, yeasticidal, fungicidal, limited virucidal action (incl. HBV, HIV, HCV), adenovirus, murine norovirus (MNV), polyoma- and rotaviruses.
In order to avoid premature drying out of the wipes, please reseal the pack immediately after use. Once opened, the wipes have a shelf life of three months.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904