Nitrile gloves, Semperguard® Nitrile Xtra Lite
Supplier: SEMPERIT TECHN PROD
816780233 816780235 816780237
SEMP816780233EA 31.7 EUR
SEMP816780233 SEMP816780235 SEMP816780237
Nitrile gloves, Semperguard® Nitrile Xtra Lite
Gloves
These ambidextrous disposable nitrile gloves protect against chemical splashes, grease, oils and other skin irritants. They are ideal for use in the processing of fatty foods.
  • Specially developed formulation and innovative production process for a wafer- thin glove
  • Excellent elasticity
  • Rolled cuffs for easier donning
  • Textured fingertips for improved grip
  • Free from powder, phthalates, softeners and allergenic latex proteins
  • Lavender coloured – suitable for areas subject to HACCP standards
  • Handy big pack (200 gloves for sizes S, M, L and 180 gloves for size XL) for optimised logistics
Length: 240 mm
Thickness: 0,14 mm
Colour: Lavender
EN 374, EN 455, Cat. III Food contact approved
Certifikace: EN 374, EN 455, Cat. III
Food contact approved
Upozornení: *double wall