Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
8019 8049 8029 8027
518-0022EA 286 EUR
518-0022 738-0111 518-0023 518-0025
Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
Microplates Filter Plates
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides, bead and chromatography resin-based applications, gross fractionation and lysate clarification, and general filtration.
  • Smooth well wall provides efficient bead recovery, ensuring reproducible results
  • High performance membrane - does not trap microspheres in the membrane matrix
  • Low levels of false positives - in serological assays, Supor® membrane effectively removes IgG complexes, thus reducing non specific reactivity of the microspheres
  • Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume; outlet tip geometry minimises sample leakage and loss during incubation steps so that acquisition times are not affected
  • Low non specific binding - intrinsic plate and membrane properties minimise target loss
Applications for AcroPrep™ Advance 96-well filter plates for multiplexing include bead-based multiplexing assays and flow cytometry.