Nucleic acid and protein purification systems, KingFisher™ mL
Supplier: Thermo Fisher Scientific
KingFisher™
735-0274EA 19540 EUR
735-0274
Nucleic acid and protein purification systems, KingFisher™ mL
Automated Nucleic Acid Purification Systems
The KingFisher™ magnetic particle processors are designed to automate time consuming sample processing of nucleic acids, proteins or cells from virtually any source (blood, cell cultures, tissue lysates, soil, faeces, for example). The KingFisher™ family consists of instruments with different throughput and volume capacities to fulfil individual sample processing needs. Each system consists of an instrument, specially designed consumables, and software. The KingFisher™ systems offer rapid and reproducible purification of high quality DNA, RNA, proteins and cells for various types of downstream applications, including genomics and proteomics, drug discovery, forensics, biomarker discovery, quality control, and veterinary assays.
  • High speed, fully automated purification
  • Purification of any sample from any source
  • Ready made and custom protocols
  • Easy operation via instrument or external PC
KingFisher™ ml is designed for easy operation up to 15 samples, processing volumes from 50 to 1000 μl in tube strips.