ESD safety shoes, lace-up, SRC, Schweinfurt, 21388, 21488
Supplier: SCHURR SCHUHVERTRIEB
2.1388.01.41 2.1388.01.42 2.1488.01.36 2.1488.01.37 2.1488.01.38 2.1488.01.39 2.1488.01.40 2.1388.01.43 2.1388.01.44 2.1388.01.45 2.1388.01.46 2.1388.01.47 2.1388.01.48 2.1488.02.36 2.1488.02.37 2.1488.02.38 2.1488.02.39 2.1488.02.40 2.1388.02.41 2.1388.02.42 2.1388.02.43 2.1388.02.44 2.1388.02.45 2.1388.02.46 2.1388.02.47 2.1388.02.48
SCRR2.1388.01.41EA 0 EUR
SCRR2.1388.01.41 SCRR2.1388.01.42 SCRR2.1488.01.36 SCRR2.1488.01.37 SCRR2.1488.01.38 SCRR2.1488.01.39 SCRR2.1488.01.40 SCRR2.1388.01.43 SCRR2.1388.01.44 SCRR2.1388.01.45 SCRR2.1388.01.46 SCRR2.1388.01.47 SCRR2.1388.01.48 SCRR2.1488.02.36 SCRR2.1488.02.37 SCRR2.1488.02.38 SCRR2.1488.02.39 SCRR2.1488.02.40 SCRR2.1388.02.41 SCRR2.1388.02.42 SCRR2.1388.02.43 SCRR2.1388.02.44 SCRR2.1388.02.45 SCRR2.1388.02.46 SCRR2.1388.02.47 SCRR2.1388.02.48
ESD safety shoes, lace-up, SRC, Schweinfurt, 21388, 21488
Shoes and Boots
ESD safety shoe made from leather, DRYtech® lining with Cambrelle® mesh insert and an anti-slip and anti-static 'A4' nitrile rubber sole; with adjustable heel strap.
  • Antibacterial and breathable DRYtech® textile lining
  • Removable insole made from PUR-foam
  • Triple shock absorber at heel and ball of the foot relieves joints
  • Protective steel toe cap (200 J)
For ESD and kitchen use.
Colour: White or black
Width: 9
Weight: 570 g
Certifikace: EN 61340 (ESD); EN ISO 20347:2012; S1, SRC