2-Cyclohexylethylboronic acid pinacol ester 96%
Supplier: Thermo Fisher Scientific
H31156.03 H31156.06
H31156.03EA 58.8 EUR
H31156.03 H31156.06
2-Cyclohexylethylboronic acid pinacol ester 96%
2-Cyclohexylethylboronic acid pinacol ester
Purity: 96% CAS No.: 167692-95-7 Molecular Formula: C14H27BO2 Formula Weight: 238.18 MDL No.: MFCD12546193
Formula: C₁₄H₂₇BO₂
MW: 238.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546193
CAS Number: 167692-95-7