Nitronium tetrafluoroborate 0.3->0.5 M in sulpholane
Supplier: Thermo Scientific
Danger
Synonyms: Nitryl tetrafluoroborate
H34069.AH H34069.AC
H34069.AHEA 398 EUR
H34069.AH H34069.AC
Nitronium tetrafluoroborate 0.3->0.5 M in sulpholane
Nitronium tetrafluoroborate
CAS No.: 13826-86-3 Molecular Formula: BF4NO2 Formula Weight: 132.81 Storage Temperature: 2°C to 8°C UN No.: 3265 MDL No.: MFCD00011432
Formula: BF₄NO₂
MW: 132.81 g/mol
Density: 1.268
Storage Temperature: Refrigerator
MDL Number: MFCD00011432
CAS Number: 13826-86-3
EINECS: 237-533-5
UN: 3265
ADR: 8,III