1H-Tetrazole 0.45 mol/l in acetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tetrazole
88185-100ML 88185-500ML
SIAL88185-100MLEA 134 EUR
SIAL88185-100ML SIAL88185-500ML
1H-Tetrazole 0.45 mol/l in acetonitrile, Sigma-Aldrich®
1H-Tetrazole
Formula: CH₂N₄
MW: 70.05 g/mol
MDL Number: MFCD00005247
CAS Number: 288-94-8
EINECS: 206-023-4