Tri(1-naphthyl)phosphine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

667218-1G 667218-5G
SIAL667218-1GEA 94.7 EUR
SIAL667218-1G SIAL667218-5G
Tri(1-naphthyl)phosphine, Sigma-Aldrich®
Tri(1-naphthyl)phosphine
Formula: C₃₀H₂₁P
MW: 412.46 g/mol
MDL Number: MFCD00046400
CAS Number: 3411-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR