1-Methyl-1H-indazole-4-boronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

725323-250MG 725323-1G
SIAL725323-250MGEA 70.2 EUR
SIAL725323-250MG SIAL725323-1G
1-Methyl-1H-indazole-4-boronic acid pinacol ester, Sigma-Aldrich®
1-Methyl-1H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MW: 258.1239 g/mol
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR