4-(Bromomethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole

Supplier: Thermo Scientific

RF00823.250 RF00823.1
MAYBRF00823.250EA 0 EUR
MAYBRF00823.250 MAYBRF00823.1
4-(Bromomethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole
4-(Bromomethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole
Formula: C₁₁H₈BrCl₂NO
MDL Number: MFCD00831541
CAS Number: 287176-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR