Okadaic acid sodium salt ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium salt, OA sodium salt
J62463.XS J62463.XG J62463.IX
J62463.XSEA 517 EUR
J62463.XS J62463.XG J62463.IX
Okadaic acid sodium salt ≥98%
Okadaic acid sodium salt
Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210208
CAS Number: 209266-80-8
UN: 3462
ADR: 6.1 I