Okadaic acid sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium salt, OA sodium salt
M-Clarity™ quality level = MQ200
O7760-10UG O7760-25UG O7760-50UG
SIALO7760-10UGEA 114 EUR
SIALO7760-10UG SIALO7760-25UG SIALO7760-50UG
Okadaic acid sodium salt, Sigma-Aldrich®
Okadaic acid sodium salt
Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
MDL Number: MFCD00210208
CAS Number: 209266-80-8