Okadaic acid ammonium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Halochondrine A ammonium salt, OA
M-Clarity™ quality level = MQ200
O8010-10UG
O8010-25UG
O8010-50UG
SIALO8010-10UGEA
256
EUR
SIALO8010-10UG
SIALO8010-25UG
SIALO8010-50UG
Okadaic acid ammonium salt, Sigma-Aldrich®
Okadaic acid ammonium salt
Formula:
C₄₄H₆₇O₁₃·NH₄ |
MDL Number:
MFCD00153856 CAS Number: 155716-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...