3-(Trimethylsilylethynyl)pyridine ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-((Trimethylsilyl)ethynyl)pyridine
H55101.03 H55101.06 H55101.14
H55101.03EA 26.3 EUR
H55101.03 H55101.06 H55101.14
3-(Trimethylsilylethynyl)pyridine ≥97%
3-(Trimethylsilylethynyl)pyridine
Formula: C₁₀H₁₃NSi
MW: 175.31 g/mol
Boiling Pt: 229 °C
Density: 0.922
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863744
CAS Number: 80673-00-3