3-Ethynylthiophene 96%
Supplier: Thermo Fisher Scientific
Warning
H55898.MD H55898.03 H55898.06
H55898.MDEA 36.2 EUR
H55898.MD H55898.03 H55898.06
3-Ethynylthiophene 96%
3-Ethynylthiophene
Formula: C₆H₄S
MW: 108.16 g/mol
Boiling Pt: 152…153 °C
Density: 1.098
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD04039973
CAS Number: 67237-53-0
UN: 1993
ADR: 3 III