Isoquinolin-1-ylmethanol

Supplier: Thermo Scientific

Synonyms: 1-(Hydroxymethyl)isoquinoline

CC05109.250 CC05109.1
MAYBCC05109.250EA 0 EUR
MAYBCC05109.250 MAYBCC05109.1
Isoquinolin-1-ylmethanol
Isoquinolin-1-ylmethanol
Formula: C₁₀H₉NO
MW: 159.1846 g/mol
MDL Number: MFCD03086097
CAS Number: 27311-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR