1-Methylindole-5-boronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
M-Clarity™ quality level = MQ100
640395-1G
640395-5G
SIAL640395-1GEA
243
EUR
SIAL640395-1G
SIAL640395-5G
1-Methylindole-5-boronic acid pinacol ester, Sigma-Aldrich®
1-Methylindole-5-boronic acid pinacol ester
Formula:
C₁₅H₂₀BNO₂ MW: 257.1358 g/mol |
MDL Number:
MFCD05663865 CAS Number: 837392-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...