Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate

Supplier: Thermo Scientific

Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

EN00291.1 EN00291EA EN00291EE
MAYBEN00291.1EA 0 EUR
MAYBEN00291.1 MAYBEN00291EA MAYBEN00291EE
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD00051652
CAS Number: 31037-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR