5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Supplier: Thermo Scientific
CD10384.250 CD10384.1
MAYBCD10384.250EA 0 EUR
MAYBCD10384.250 MAYBCD10384.1
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Formula: C₅H₈ClN₃O₂S
MW: 209.66 g/mol
MDL Number: MFCD00105874
CAS Number: 88398-46-3