5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonic acid

Supplier: Thermo Scientific

CD10444.250 CD10444.1
MAYBCD10444.250EA 0 EUR
MAYBCD10444.250 MAYBCD10444.1
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonic acid
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonic acid
Formula: C₅H₇ClN₂O₃S
CAS Number: 499785-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR