1H-Indazole-5-boronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

M-Clarity™ quality level = MQ100

692727-1G 692727-5G
SIAL692727-1GEA 117 EUR
SIAL692727-1G SIAL692727-5G
1H-Indazole-5-boronic acid pinacol ester, Sigma-Aldrich®
1H-Indazole-5-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
MDL Number: MFCD07367523
CAS Number: 862723-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR