3-Amino-5-iodo-1H-indazole 97%
Supplier: Thermo Scientific
Warning
H34227.03 H34227.06
H34227.03EA 67.6 EUR
H34227.03 H34227.06
3-Amino-5-iodo-1H-indazole 97%
3-Amino-5-iodo-1H-indazole
Formula: C₇H₆IN₃
MW: 259.05 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD09261138
CAS Number: 88805-76-9