3-(2,4-Dichlorophenyl)isoxazole

Supplier: Thermo Scientific

SEW05782.1 SEW05782.10 SEW05782.50
MAYBSEW05782.1EA 0 EUR
MAYBSEW05782.1 MAYBSEW05782.10 MAYBSEW05782.50
3-(2,4-Dichlorophenyl)isoxazole
3-(2,4-Dichlorophenyl)isoxazole
Formula: C₉H₅Cl₂NO
CAS Number: 260973-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR