N,N-Dimethyl-1-dodecylamine N-oxide ∼30% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
40236-1L 40236-250ML
SIAL40236-1LEA 194 EUR
SIAL40236-1L SIAL40236-250ML
N,N-Dimethyl-1-dodecylamine N-oxide ∼30% in aqueous solution, Sigma-Aldrich®
N,N-Dimethyl-1-dodecylamine N-oxide
Formula: C₁₄H₃₁NO
MDL Number: MFCD00002049
CAS Number: 1643-20-5
EINECS: 216-700-6