N,N-Dimethyl-1-dodecylamine N-oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-ethyl-N-oxido-dodecan-1-amine, LDAO, Lauryldimethylamine oxide, N,N-Dimethyldodecan-1-amine oxide, Lauramine oxide, N,N-Dimethyl-N-dodecylamine N-oxide
40234-25G 40234-5G
SIAL40234-25GEA 827 EUR
SIAL40234-25G SIAL40234-5G
N,N-Dimethyl-1-dodecylamine N-oxide, Sigma-Aldrich®
N,N-Dimethyl-1-dodecylamine N-oxide
Formula: C₁₄H₃₁NO
MW: 229.40 g/mol
MDL Number: MFCD00002049
CAS Number: 1643-20-5
EINECS: 216-700-6