2-Phenylethylboronic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Phenethylboronic acid
M-Clarity™ quality level = MQ100
588423-10G
588423-1G
SIAL588423-10GEA
263
EUR
SIAL588423-10G
SIAL588423-1G
2-Phenylethylboronic acid, Sigma-Aldrich®
2-Phenylethylboronic acid
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol |
MDL Number:
MFCD01631226 CAS Number: 34420-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...