α-Bromo-4-chlorophenylacetic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

638676-25G 638676-5G
SIAL638676-25GEA 823 EUR
SIAL638676-25G SIAL638676-5G
α-Bromo-4-chlorophenylacetic acid, Sigma-Aldrich®
α-Bromo-4-chlorophenylacetic acid
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD08276760
CAS Number: 3381-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR