2-Chloro-N-(2,3-dimethylphenyl)benzamide ≥97%
Supplier: Thermo Scientific
Warning
H56225.MD H56225.03
H56225.MDEA 201 EUR
H56225.MD H56225.03
2-Chloro-N-(2,3-dimethylphenyl)benzamide ≥97%
2-Chloro-N-(2,3-dimethylphenyl)benzamide
Formula: C₁₅H₁₄ClNO
MW: 259.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018254
CAS Number: 196617-88-6