Probenecid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-[(dipropylamino)sulphonyl]benzoic acid
M-Clarity™ quality level = MQ200
P8761-100G
P8761-25G
SIALP8761-100GEA
399
EUR
SIALP8761-100G
SIALP8761-25G
Probenecid, Sigma-Aldrich®
Probenecid
Formula:
C₁₃H₁₉NO₄S MW: 285.4 g/mol |
MDL Number:
MFCD00038402 CAS Number: 57-66-9 EINECS: 200-344-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...