1-(Methylsulphonyl)-4-nitrobenzene
Supplier: Thermo Scientific
RH00770DA
RH00770EA
RH00770.50
MAYBRH00770DAEA
0
EUR
MAYBRH00770DA
MAYBRH00770EA
MAYBRH00770.50
1-(Methylsulphonyl)-4-nitrobenzene
1-(Methylsulphonyl)-4-nitrobenzene
Formula:
C₇H₇NO₄S MW: 201.20 g/mol |
MDL Number:
MFCD00047803 CAS Number: 2976-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...