1,2-Dimethoxy-4,5-dinitrobenzene 96%
Supplier: Thermo Scientific
|
Danger
|
L11706.03
L11706.06
L11706.03EA
58.7
EUR
L11706.03
L11706.06
1,2-Dimethoxy-4,5-dinitrobenzene 96%
1,2-Dimethoxy-4,5-dinitrobenzene
Purity: 96%
CAS No.: 3395-03-7
Molecular Formula: C8H8N2O6
Formula Weight: 228.16
Melting Point: 131-134°
Beilstein Reference: 2055812
UN No.: 2811
MDL No.: MFCD00082695
Formula:
C₈H₈N₂O₆ MW: 228.16 g/mol Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082695 CAS Number: 3395-03-7 ADR: 6.1,III |