2,4-Dimethoxybenzenesulphonamide 96%
Supplier: Thermo Fisher Scientific
Warning
H31707.03 H31707.06
H31707.03EA 52.8 EUR
H31707.03 H31707.06
2,4-Dimethoxybenzenesulphonamide 96%
2,4-Dimethoxybenzenesulphonamide
Purity: 96% CAS No.: 51770-71-9 Molecular Formula: C8H11NO4S Formula Weight: 217.25 Melting Point: 166-170° MDL No.: MFCD06147001
Formula: C₈H₁₁NO₄S
MW: 217.25 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06147001
CAS Number: 51770-71-9