Methyl-2-iodo-5-methylbenzoate 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Methyl 2-Iodo-5-methylbenzoate
H33853.03 H33853.06
H33853.03EA 34.2 EUR
H33853.03 H33853.06
Methyl-2-iodo-5-methylbenzoate 95%
Methyl 2-iodo-5-methylbenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198129
CAS Number: 103440-52-4