Isoquinoline-1-carbonitrile

Supplier: Thermo Scientific

Synonyms: 1-Isoquinolinecarbonitrile

BTB09817.1 BTB09817.10
MAYBBTB09817.1EA 0 EUR
MAYBBTB09817.1 MAYBBTB09817.10
Isoquinoline-1-carbonitrile
Isoquinoline-1-carbonitrile
Formula: C₁₀H₆N₂
MW: 154.17 g/mol
MDL Number: MFCD00134166
CAS Number: 1198-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR