2-Hydroxy-6-methylnicotinonitrile ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 6-Methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-Cyano-6-methyl-2-pyridone
B20188.09 B20188.18 B20188.30
B20188.09EA 69.9 EUR
B20188.09 B20188.18 B20188.30
2-Hydroxy-6-methylnicotinonitrile ≥98%
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Purity: 98% CAS No.: 4241-27-4 Molecular Formula: C7H6N2O Formula Weight: 134.14 Melting Point: ca 298° dec. Beilstein Reference: 120412
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: ∼ 298 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006013
CAS Number: 4241-27-4
EINECS: 224-202-5