Potassium nitrate-[¹⁵N] (98% ¹⁵N), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
335134-1G 335134-250MG
SIAL335134-1GEA 155 EUR
SIAL335134-1G SIAL335134-250MG
Potassium nitrate-[¹⁵N] (98% ¹⁵N), Sigma-Aldrich®
Formula: KNO₃
Storage Temperature: Ambient
MDL Number: MFCD00075049
CAS Number: 57654-83-8