3-(Chloromethyl)benzoic acid, Sigma-Aldrich®

Supplier: Merck


MQ100

424315-25G 424315-5G
SIAL424315-25GEA 115 EUR
SIAL424315-25G SIAL424315-5G
3-(Chloromethyl)benzoic acid, Sigma-Aldrich®
3-(Chloromethyl)benzoic acid
Formula: C₈H₇ClO₂
MW: 170.59 g/mol
MDL Number: MFCD00191922
CAS Number: 31719-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR