4-(Chloromethyl)benzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
270792-10G 270792-50G
SIAL270792-10GEA 62 EUR
SIAL270792-10G SIAL270792-50G
4-(Chloromethyl)benzoic acid, Sigma-Aldrich®
4-(Chloromethyl)benzoic acid
Formula: C₈H₇ClO₂
MDL Number: MFCD00002568
CAS Number: 1642-81-5
EINECS: 216-697-1