4-tert-Butylbenzamide
Supplier: Thermo Scientific
BTB08864.1
BTB08864.10
BTB08864.50
MAYBBTB08864.1EA
0
EUR
MAYBBTB08864.1
MAYBBTB08864.10
MAYBBTB08864.50
4-tert-Butylbenzamide
4-tert-Butylbenzamide
Formula:
C₁₁H₁₅NO MW: 177.25 g/mol |
MDL Number:
MFCD00017139 CAS Number: 56108-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...