3,5-Dibromo-4-methoxybenzonitrile
Supplier: Thermo Scientific
Synonyms: Bromoxynil Methyl Ether
SPB00194.1 SPB00194.10
MAYBSPB00194.1EA 0 EUR
MAYBSPB00194.1 MAYBSPB00194.10
3,5-Dibromo-4-methoxybenzonitrile
3,5-Dibromo-4-methoxybenzonitrile
Formula: C₈H₅Br₂NO
MW: 290.939 g/mol
MDL Number: MFCD00017333
CAS Number: 3336-39-8