N-(2-Bromoethyl)-3-(trifluoromethyl)benzenesulphonamide

Supplier: Thermo Scientific

BTB09485.250 BTB09485.1
MAYBBTB09485.250EA 0 EUR
MAYBBTB09485.250 MAYBBTB09485.1
N-(2-Bromoethyl)-3-(trifluoromethyl)benzenesulphonamide
N-(2-Bromoethyl)-3-(trifluoromethyl)benzenesulphonamide
Formula: C₉H₉BrF₃NO₂S
MDL Number: MFCD00829016
CAS Number: 246236-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR