1,3,5-Trifluoro-2,4,6-triiodobenzene ≥97%
Supplier: Thermo Scientific
Warning
H33116.03 H33116.06
H33116.03EA 120 EUR
H33116.03 H33116.06
1,3,5-Trifluoro-2,4,6-triiodobenzene ≥97%
1,3,5-Trifluoro-2,4,6-triiodobenzene
Formula: C₆F₃I₃
MW: 509.77 g/mol
Melting Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD06248899
CAS Number: 84322-56-5