2-(Methylthio)benzonitrile
Supplier: Thermo Scientific
TL00713.1
TL00713.10
TL00713.50
MAYBTL00713.1EA
0
EUR
MAYBTL00713.1
MAYBTL00713.10
MAYBTL00713.50
2-(Methylthio)benzonitrile
2-(Methylthio)benzonitrile
Formula:
C₈H₇NS MW: 149.22 g/mol |
MDL Number:
MFCD00015557 CAS Number: 6609-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...