1-(2-Aminoethyl)piperidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-(2-Aminoethyl)piperidine

M-Clarity™ quality level = MQ100

141666-10G 141666-5G
SIAL141666-10GEA 84.5 EUR
SIAL141666-10G SIAL141666-5G
1-(2-Aminoethyl)piperidine, Sigma-Aldrich®
1-(2-Aminoethyl)piperidine
Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD00006516
CAS Number: 27578-60-5
EINECS: 248-540-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR