Cerium(IV) oxide 25% in water, colloidal dispersion, NanoArc® CE-6440
Supplier: Thermo Scientific
Warning
Synonyms: Ceric oxide, Cerium dioxide
46763.22 46763.36
46763.22EA 66.6 EUR
46763.22 46763.36
Cerium(IV) oxide 25% in water, colloidal dispersion, NanoArc® CE-6440
Cerium (IV) oxide
CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 30nm APS for dry powder (typical) Odour: Odourless MDL No.: MFCD00010933
Suitable for use in ultrafine acid side polishing slurries
Formula: CeO₂
MW: 172.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4